Assay Details
Target Gene Details
Entrez Gene ID: | 381673 |
Gene Name: | RIKEN cDNA A330070K13 gene |
Gene Aliases: |
Gm1051 |
Location: |
Chr.5:130378851-130384631 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| A330070K13Rik | NM_198665.1 | NP_941067.1 | ||
| AK039605.1 | BAC30397.1 |
Target Gene Details
Entrez Gene ID: | 140904 |
Gene Name: | calneuron 1 |
Gene Aliases: |
9630012C17Rik, Cabp8, MNCb-0849 |
Location: |
Chr.5:130369356-130846325 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Caln1 | NM_181045.1 | NP_851388.1 | ||
| AF282251.1 | AAK15156.1 | |||
| AK035865.1 | BAC29219.1 | |||
| BY127416.1 |
More Information
Set Membership: |
|

Genomic Map