Assay Details
Target Gene Details
Entrez Gene ID: | 14570 |
Gene Name: | Rho GDP dissociation inhibitor (GDI) gamma |
Gene Aliases: |
Gdi5, RIP2, Rho-GDI-3, Rho-GDI2 |
Location: |
Chr.17:26199183-26201350 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Arhgdig | NM_008113.3 | NP_032139.1 | ||
| AK144321.1 | BAE25831.1 | |||
| BC002032.1 | AAH02032.1 | |||
| BC158003.1 | ||||
| L42463.1 | AAC37704.1 | |||
| U73198.1 | AAB18196.1 |
More Information
Set Membership: |
|

Genomic Map