Assay Details
Target Gene Details
Entrez Gene ID: | 70843 |
Gene Name: | keratin 28 |
Gene Aliases: |
4733401L19Rik, Krt25d |
Location: |
Chr.11:99364861-99379930 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Krt28 | NM_027574.1 | NP_081850.1 | ||
| AB288231.1 | ||||
| AK014642.1 | BAB29485.1 |
More Information
Set Membership: |
|

Genomic Map