Assay Details
Target Gene Details
Entrez Gene ID: | 237934 |
Gene Name: | keratin 39 |
Gene Aliases: |
4732494G06Rik, Ka35 |
Location: |
Chr.11:99514293-99521338 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Krt39 | NM_213730.2 | NP_998895.2 | ||
| BC119541.1 | ||||
| BC128563.1 |
More Information
Set Membership: |
|

Genomic Map