Assay Details
Target Gene Details
Entrez Gene ID: | 238076 |
Gene Name: | potassium voltage-gated channel, delayed-rectifier, subfamily S, member 3 |
Gene Aliases: |
- |
Location: |
Chr.12:11090202-11158735 on Build GRCm38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Kcns3 | NM_001168564.1 | NP_001162036.1 | ||
| NM_173417.3 | NP_775593.1 | |||
| AK046054.1 | BAC32583.1 | |||
| AK162957.1 | BAE37134.1 | |||
| BC132164.1 | ||||
| BC132464.1 |
More Information
Set Membership: |
|

Genomic Map