Assay Details
Target Gene Details
Entrez Gene ID: | 27356 |
Gene Name: | insulin-like 6 |
Gene Aliases: |
RIF1 |
Location: |
Chr.19:29321354-29325318 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Insl6 | NM_013754.1 | NP_038782.1 | ||
| AF135823.1 | ||||
| AF143807.2 | AAD30158.2 | |||
| AK005622.1 | BAB24156.1 | |||
| BC028752.1 | AAH28752.1 |
More Information
Set Membership: |
|

Genomic Map