Assay Details
Target Gene Details
Entrez Gene ID: | 71760 |
Gene Name: | ethanolamine phosphate phospholyase |
Gene Aliases: |
1300019H02Rik, AI195447, Agxt2l1 |
Location: |
Chr.3:130617448-130635750 on Build GRCm38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Etnppl | NM_001163587.1 | NP_001157059.1 | ||
| NM_027907.3 | NP_082183.2 | |||
| AK005060.1 | BAB23784.1 | |||
| AK049937.1 | BAC33993.1 | |||
| BC043680.1 | AAH43680.2 | |||
| BC058592.1 | AAH58592.1 |
More Information
Set Membership: |
|

Genomic Map