Assay Details
Target Gene Details
Entrez Gene ID: | 75732 |
Gene Name: | IQ motif containing D |
Gene Aliases: |
4933433C09Rik, AI662646 |
Location: |
Chr.5:120586060-120607243 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Iqcd | NM_029408.2 | NP_083684.1 | ||
| AK017032.1 | BAB30561.1 | |||
| AK076996.1 | BAC36556.1 | |||
| BC100441.1 | AAI00442.1 | |||
| BC125429.1 | ||||
| BC137644.1 |
More Information
Set Membership: |
|

Genomic Map