Assay Details
Target Gene Details
Entrez Gene ID: | 329581 |
Gene Name: | baculoviral IAP repeat-containing 7 (livin) |
Gene Aliases: |
E130019N06, KIAP, Livin, ML-IAP |
Location: |
Chr.2:180928152-180935028 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Birc7 | NM_001163247.1 | NP_001156719.1 | ||
| AK053463.1 | ||||
| BC107260.2 | ||||
| BC125013.1 |
More Information
Set Membership: |
|

Genomic Map