Assay Details
Target Gene Details
Entrez Gene ID: | 12869 |
Gene Name: | cytochrome c oxidase subunit VIIIb |
Gene Aliases: |
Cox8h, CoxVIII-H |
Location: |
Chr.7:140898942-140900446 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cox8b | NM_007751.3 | NP_031777.1 | ||
| AA028501.1 | ||||
| AA511205.1 | ||||
| AK003116.1 | BAB22576.1 | |||
| BC027531.1 | AAH27531.1 | |||
| BC086930.1 | AAH86930.1 | |||
| BE626379.1 | ||||
| U15541.1 | AAA80280.1 |
More Information
Set Membership: |
|

Genomic Map