Assay Details
Target Gene Details
Entrez Gene ID: | 66067 |
Gene Name: | GTP-binding protein 8 (putative) |
Gene Aliases: |
0610037H22Rik |
Location: |
Chr.16:44738879-44746377 on Build GRCm38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gtpbp8 | NM_001159329.1 | NP_001152801.1 | ||
| NM_025332.3 | NP_079608.1 | |||
| AK002771.1 | ||||
| AK010979.1 | BAB27301.1 | |||
| AK147840.1 | BAE28175.1 | |||
| BC107216.1 | AAI07217.1 | |||
| BC107217.1 | AAI07218.1 |
More Information
Set Membership: |
|

Genomic Map