Assay Details
Target Gene Details
Entrez Gene ID: | 75465 |
Gene Name: | dynein light chain roadblock-type 2 |
Gene Aliases: |
1700009A04Rik, DNLC2B, Dncl2b |
Location: |
Chr.8:116505015-116515915 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dynlrb2 | NM_029297.1 | NP_083573.1 | ||
| AK005789.1 | BAB24240.1 | |||
| BC048623.1 | AAH48623.1 |
More Information
Set Membership: |
|

Genomic Map