Assay Details
Target Gene Details
Entrez Gene ID: | 260409 |
Gene Name: | CDC42 effector protein (Rho GTPase binding) 3 |
Gene Aliases: |
3200001F04Rik, AA986861, Borg2, Cep3, UB1 |
Location: |
Chr.17:79333964-79355091 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cdc42ep3 | NM_026514.2 | NP_080790.1 | ||
| AK012309.1 | BAB28155.1 | |||
| AK014281.1 | BAB29241.1 | |||
| AK019447.1 | BAB31723.1 | |||
| AK076782.1 | BAC36479.1 | |||
| AK150214.1 | BAE29384.1 | |||
| BC034714.1 | AAH34714.1 | |||
| BC092297.1 | AAH92297.1 |
More Information
Set Membership: |
|

Genomic Map