Assay Details
Target Gene Details
Entrez Gene ID: | 68644 |
Gene Name: | abhydrolase domain containing 14A |
Gene Aliases: |
1110013B16Rik, AW558221, Dorz1 |
Location: |
Chr.9:106440051-106451384 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Abhd14a | NM_001110271.1 | NP_001103741.1 | ||
| NM_001110272.1 | NP_001103742.1 | |||
| NM_145919.2 | NP_666031.2 | |||
| AB073619.1 | BAB91136.1 | |||
| AK003653.1 | ||||
| BC006879.1 | AAH06879.1 | |||
| BY144149.1 | ||||
| CB952955.1 |
More Information
Set Membership: |
|

Genomic Map