Assay Details
Target Gene Details
Entrez Gene ID: | 69327 |
Gene Name: | RIKEN cDNA 1700007K13 gene |
Gene Aliases: |
Rbest47 |
Location: |
Chr.2:28462001-28466324 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 1700007K13Rik | NM_027040.1 | NP_081316.1 | ||
| AK005731.1 | BAB24210.1 | |||
| AY927869.1 | AAX22218.1 | |||
| BC099566.1 | AAH99566.1 |
More Information
Set Membership: |
|

Genomic Map