Assay Details
Target Gene Details
Entrez Gene ID: | 68099 |
Gene Name: | family with sequence similarity 92, member A |
Gene Aliases: |
6720467C03Rik, Fam92a1 |
Location: |
Chr.4:12153722-12172022 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam92a | NM_026558.4 | NP_080834.3 | ||
| AK013129.1 | ||||
| AK019574.1 | BAB31798.1 | |||
| AK032690.1 | ||||
| AK032976.1 | 1 | 1037 | ||
| AK078438.1 | BAC37273.1 | |||
| AK088705.1 | BAC40516.1 | |||
| BC055848.1 | 2 | 550 | AAH55848.1 |
More Information
Set Membership: |
|

Genomic Map