Assay Details
Target Gene Details
Entrez Gene ID: | 100126226 |
Gene Name: | RIKEN cDNA 5430421N21 gene |
Gene Aliases: |
Krt83 |
Location: |
Chr.15:101485130-101491512 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 5430421N21Rik | NM_001201323.1 | NP_001188252.1 | ||
| AK133718.1 | ||||
| M92088.1 | AAA39273.1 |
More Information
Set Membership: |
|

Genomic Map