Assay Details
Target Gene Details
Entrez Gene ID: | 56752 |
Gene Name: | aldehyde dehydrogenase 9, subfamily A1 |
Gene Aliases: |
AA139417, Abaldh, ESTM40, TMABA-DH, Tmabadh |
Location: |
Chr.1:167349991-167368530 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Aldh9a1 | NM_019993.3 | NP_064377.2 | ||
| AB296386.1 | ||||
| AF170919.1 | AAF43599.1 | |||
| AK132336.1 | BAE21113.1 | |||
| AK149589.1 | BAE28979.1 | |||
| AK150093.1 | BAE29301.1 | |||
| AK154914.1 | BAE32922.1 | |||
| AK162354.1 | BAE36870.1 | |||
| AK165730.1 | BAE38357.1 | |||
| AK168261.1 | BAE40209.1 | |||
| AK168309.1 | BAE40249.1 | |||
| AK168878.1 | BAE40696.1 | |||
| BC003297.1 | AAH03297.1 |
More Information
Set Membership: |
|

Genomic Map