Assay Details
Target Gene Details
Entrez Gene ID: | 22144 |
Gene Name: | tubulin, alpha 3A |
Gene Aliases: |
AI528779, M[a]3, Tuba3 |
Location: |
Chr.6:125278274-125286042 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tuba3a | NM_009446.2 | NP_033472.1 | ||
| AK132775.1 | BAE21352.1 | |||
| BC050770.1 | AAH50770.1 | |||
| BC089547.1 | AAH89547.1 | |||
| M13442.1 | AAA40501.1 |
More Information
Set Membership: |
|

Genomic Map