Assay Details
Target Gene Details
Entrez Gene ID: | 225058 |
Gene Name: | predicted gene 4832 |
Gene Aliases: |
EG225058 |
Location: |
Chr.17:88066125-88131186 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gm4832 | NM_001190356.1 | NP_001177285.1 | ||
| AK139504.1 | BAE24039.1 |
More Information
Set Membership: |
|

Genomic Map