Assay Details
Target Gene Details
Entrez Gene ID: | 407819 |
Gene Name: | cDNA sequence BC031181 |
Gene Aliases: |
- |
Location: |
Chr.18:75005274-75009934 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BC031181 | NM_001001181.3 | NP_001001181.1 | ||
| AK031999.1 | BAE20478.1 | |||
| AK077642.1 | BAC36920.1 | |||
| AK131658.1 | BAE20746.1 | |||
| AK182472.1 | ||||
| AK216918.1 | ||||
| BC016084.1 | AAH16084.1 | |||
| BC027819.1 | AAH27819.1 | |||
| BC031181.1 | AAH31181.1 |
More Information
Set Membership: |
|

Genomic Map