Assay Details
Target Gene Details
Entrez Gene ID: | 67477 |
Gene Name: | abhydrolase domain containing 15 |
Gene Aliases: |
1300007F04Rik |
Location: |
Chr.11:77515117-77520628 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Abhd15 | NM_026185.4 | NP_080461.3 | ||
| AK004929.1 | BAC25102.1 | |||
| AK018696.1 | BAB31349.1 | |||
| AK042032.1 | BAC31138.1 | |||
| AK046590.1 | BAE20655.1 | |||
| AK153809.1 | ||||
| FJ154119.1 |
More Information
Set Membership: |
|

Genomic Map