Assay Details
Target Gene Details
Entrez Gene ID: | 66934 |
Gene Name: | DSN1, MIND kinetochore complex component, homolog (S. cerevisiae) |
Gene Aliases: |
1700022L09Rik, AW552447 |
Location: |
Chr.2:156995062-157007075 on Build GRCm38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dsn1 | NM_025853.3 | NP_080129.2 | ||
| AK006246.1 | BAB24479.1 | |||
| AK017740.1 | BAB30906.1 | |||
| AK017813.1 | BAB30950.1 | |||
| AK139887.1 | ||||
| AK145373.1 | ||||
| BC025079.1 | AAH25079.1 | |||
| BC046807.1 | AAH46807.1 |
More Information
Set Membership: |
|

Genomic Map