Assay Details
Target Gene Details
Entrez Gene ID: | 239833 |
Gene Name: | leishmanolysin-like (metallopeptidase M8 family) |
Gene Aliases: |
5330415H22Rik, AI465495 |
Location: |
Chr.16:33061871-33127695 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lmln | NM_172823.2 | NP_766411.1 | ||
| AK030463.1 | BAC26972.1 | |||
| BC111899.1 | ||||
| BC112428.1 |
More Information
Set Membership: |
|

Genomic Map