Assay Details
Target Gene Details
Entrez Gene ID: | 327747 |
Gene Name: | methyltransferase like 24 |
Gene Aliases: |
9030224M15Rik |
Location: |
Chr.10:40682299-40811086 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mettl24 | NM_177793.3 | NP_808461.1 | ||
| AK033488.1 | BAC28315.1 | |||
| BC147316.1 | ||||
| BC147317.1 |
More Information
Set Membership: |
|

Genomic Map