Assay Details
Target Gene Details
Entrez Gene ID: | 57764 |
Gene Name: | netrin 4 |
Gene Aliases: |
- |
Location: |
Chr.10:93641049-93745972 on Build GRCm38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ntn4 | NM_021320.3 | NP_067295.2 | ||
| AF268066.1 | AAF91404.1 | |||
| AF281278.1 | AAG30823.1 | |||
| BC119182.1 | ||||
| BC119184.1 | ||||
| CX205916.1 |
More Information
Set Membership: |
|

Genomic Map