Assay Details
Target Gene Details
Entrez Gene ID: | 114774 |
Gene Name: | PRKC, apoptosis, WT1, regulator |
Gene Aliases: |
2310001G03Rik, PAR4, Par-4 |
Location: |
Chr.10:108332189-108414391 on Build GRCm38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pawr | NM_054056.2 | NP_473397.1 | ||
| AF377871.1 | AAK55414.1 | |||
| AW123400.1 | ||||
| DQ363525.1 | ABC96645.1 | |||
| DQ449073.1 | ||||
| DQ449074.1 |
More Information
Set Membership: |
|

Genomic Map