Assay Details
Target Gene Details
Entrez Gene ID: | 70810 |
Gene Name: | keratin 25 |
Gene Aliases: |
4631426H08Rik, Ka38, Krt25a, mIRSa1 |
Location: |
Chr.11:99315516-99322941 on Build GRCm38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Krt25 | NM_133730.1 | NP_598491.1 | ||
| AK014538.1 | ||||
| BC018391.1 | AAH18391.1 |
More Information
Set Membership: |
|

Genomic Map