Assay Details
Target Gene Details
Entrez Gene ID: | 545279 |
Gene Name: | membrane-spanning 4-domains, subfamily A, member 15 |
Gene Aliases: |
E530011F12Rik, Gm1286 |
Location: |
Chr.19:10978303-10993306 on Build GRCm38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ms4a15 | NM_001034898.2 | NP_001030070.1 | ||
| AK143446.1 | BAE25379.1 | |||
| BC147202.1 | ||||
| BC172010.1 |
More Information
Set Membership: |
|

Genomic Map