Assay Details
Target Gene Details
Entrez Gene ID: | 67553 |
Gene Name: | glutathione S-transferase, C-terminal domain containing |
Gene Aliases: |
4933434L15Rik |
Location: |
Chr.3:132981752-133091971 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gstcd | NM_026231.2 | NP_080507.2 | ||
| AK029724.1 | ||||
| AK033409.1 | ||||
| AK037169.1 | BAC29731.1 | |||
| AK044441.1 | BAC31921.1 | |||
| BC047136.1 | AAH47136.1 | |||
| BC061240.1 | AAH61240.1 |
More Information
Set Membership: |
|

Genomic Map