Assay Details
Target Gene Details
Entrez Gene ID: | 12928 |
Gene Name: | v-crk sarcoma virus CT10 oncogene homolog (avian) |
Gene Aliases: |
Crk-I, Crk-II, Crk-III, Crk3, CrkIII, Crko, c-Crk, p38 |
Location: |
Chr.11:75679248-75708428 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Crk | NM_001277219.1 | NP_001264148.1 | ||
| NM_001277221.1 | NP_001264150.1 | |||
| NM_133656.5 | NP_598417.2 | |||
| AF239673.1 | AAL27153.1 | |||
| AK028179.1 | BAC25792.1 | |||
| AK028488.1 | BAC25976.1 | |||
| AK036905.1 | BAC29633.1 | |||
| AK052795.1 | BAC35149.1 | |||
| AK076097.1 | BAC36180.1 | |||
| AK088519.1 | BAC40399.1 | |||
| AK139089.1 | BAE23885.1 | |||
| AK152549.1 | ||||
| AK154987.1 | BAE32976.1 | |||
| AK161356.1 | BAE36345.1 | |||
| AK161761.1 | BAE36562.1 | |||
| AK163291.1 | BAE37279.1 | |||
| BC012216.1 | AAH12216.1 | |||
| BC031149.1 | AAH31149.1 | |||
| S72408.1 | AAB30755.1 |
More Information
Set Membership: |
|

Genomic Map