Assay Details
Target Gene Details
Entrez Gene ID: | 75368 |
Gene Name: | RIKEN cDNA 4930558K02 gene |
Gene Aliases: |
- |
Location: |
Chr.1:161942089-161982371 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 4930558K02Rik | NM_001204904.1 | NP_001191833.1 | ||
| CF105154.1 |
Target Gene Details
Entrez Gene ID: | 67292 |
Gene Name: | phosphatidylinositol glycan anchor biosynthesis, class C |
Gene Aliases: |
3110030E07Rik, AW212108 |
Location: |
Chr.1:161969188-161973435 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pigc | NM_001039045.1 | NP_001034134.1 | ||
| NM_026078.2 | NP_080354.1 | |||
| AK008524.1 | ||||
| AK014096.1 | BAB29152.1 | |||
| AK016394.1 | ||||
| AK152660.1 | BAE31396.1 | |||
| AK155971.1 | 1 | 560 | BAE33529.1 | |
| AK161117.1 | ||||
| AK170153.1 | BAE41600.1 | |||
| BC006938.1 | AAH06938.1 |
More Information
Set Membership: |
|

Genomic Map