Assay Details
Target Gene Details
Entrez Gene ID: | 378702 |
Gene Name: | small EDRK-rich factor 2 |
Gene Aliases: |
C80083, Msmac1l, m4F5rel |
Location: |
Chr.2:121449198-121453420 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Serf2 | NM_001290837.1 | NP_001277766.1 | ||
| NM_011354.3 | NP_035484.1 | |||
| NR_110986.1 | ||||
| AA423654.1 | ||||
| AF073297.1 | AAC63515.1 | |||
| AK002351.1 | BAB22030.3 | |||
| AK008802.1 | ||||
| AK010749.1 | BAB27157.1 | |||
| AK012395.1 | ||||
| AK012603.1 | ||||
| AK018746.1 | ||||
| AK132746.1 | ||||
| AK151362.1 | BAE30337.1 | |||
| AK168858.1 | BAE40679.1 | |||
| AK169106.1 | BAE40888.1 | |||
| BC092136.1 | AAH92136.1 | |||
| BY070249.1 | ||||
| CJ073462.1 |
More Information
Set Membership: |
|

Genomic Map