Assay Details
Target Gene Details
Entrez Gene ID: | 12587 |
Gene Name: | melanoma inhibitory activity |
Gene Aliases: |
CD-RAP, Cdrap, Mia1 |
Location: |
Chr.7:27179741-27181149 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mia | NM_019394.3 | NP_062267.2 | ||
| AK135151.1 | 2 | 446 | BAE22443.1 | |
| AW047492.1 | ||||
| BE864464.1 | ||||
| CK794645.1 | ||||
| X94322.1 | CAA63983.1 |
More Information
Set Membership: |
|

Genomic Map