Assay Details
Target Gene Details
Entrez Gene ID: | 14308 |
Gene Name: | follicle stimulating hormone beta |
Gene Aliases: |
FSH, Fshbeta |
Location: |
Chr.2:107056140-107059651 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fshb | NM_008045.2 | NP_032071.1 | ||
| AK017593.1 | BAB30827.1 | |||
| BC061159.1 | AAH61159.1 |
More Information
Set Membership: |
|

Genomic Map