Assay Details
Target Gene Details
Entrez Gene ID: | 18598 |
Gene Name: | pyruvate dehydrogenase E1 alpha 2 |
Gene Aliases: |
Pdhal |
Location: |
Chr.3:141210004-141212355 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pdha2 | NM_008811.2 | 1 | 187 | NP_032837.1 |
| M76728.1 | 1 | 185 | AAA53047.1 |
More Information
Set Membership: |
|

Genomic Map