Assay Details
Target Gene Details
Entrez Gene ID: | 68046 |
Gene Name: | RIKEN cDNA 2700062C07 gene |
Gene Aliases: |
AI195775, C87515 |
Location: |
Chr.18:24470871-24477767 on Build GRCm38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 2700062C07Rik | NM_026529.4 | NP_080805.1 | ||
| AK011228.1 | ||||
| AK012467.1 | ||||
| AK013030.1 | BAB28609.1 | |||
| AK018391.1 | BAB31191.1 | |||
| AK164298.1 | ||||
| BC037075.1 | AAH37075.1 | |||
| BC084681.1 | AAH84681.1 |
More Information
Set Membership: |
|

Genomic Map