Assay Details
Target Gene Details
Entrez Gene ID: | 11609 |
Gene Name: | angiotensin II receptor, type 2 |
Gene Aliases: |
AI316812, AW107640 |
Location: |
Chr.X:21484549-21488833 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Agtr2 | NM_007429.5 | NP_031455.1 | ||
| AK086334.1 | BAC39650.1 | |||
| AK140941.1 | BAE24525.1 | |||
| L32840.1 | AAB49539.1 | |||
| S67465.1 | AAB29336.1 | |||
| U04828.1 | AAC52128.1 |
More Information
Set Membership: |
|

Genomic Map