Assay Details
Target Gene Details
Entrez Gene ID: | 219026 |
Gene Name: | epididymal protein 3B |
Gene Aliases: |
Fam12, Gm75 |
Location: |
Chr.14:51114370-51117815 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Eddm3b | NM_203508.1 | NP_987104.1 | ||
| AK136724.1 | BAE23108.1 | |||
| BC031737.1 | AAH31737.1 |
More Information
Set Membership: |
|

Genomic Map