Assay Details
Target Gene Details
Entrez Gene ID: | 15434 |
Gene Name: | homeobox D3 |
Gene Aliases: |
Hox-4.1, Hox-5.5 |
Location: |
Chr.2:74711993-74748271 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hoxd3 | NM_010468.2 | NP_034598.2 | ||
| AK133705.1 | BAE21791.1 | |||
| AK138525.1 | BAE23694.1 | |||
| AK140904.1 | 1 | 223 | ||
| BC137716.1 | ||||
| BC137718.1 | ||||
| U56400.1 | AAC52779.1 | |||
| X73573.1 | CAA51975.1 |
More Information
Set Membership: |
|

Genomic Map