Assay Details
Target Gene Details
Entrez Gene ID: | 238555 |
Gene Name: | butyrophilin, subfamily 2, member A2 |
Gene Aliases: |
Btn2, D030063K05 |
Location: |
Chr.13:23477049-23488858 on Build GRCm38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Btn2a2 | NM_001289614.1 | NP_001276543.1 | ||
| NM_001289615.1 | NP_001276544.1 | |||
| NM_175938.3 | NP_787952.2 | |||
| AK051061.1 | ||||
| AY177686.1 | AAO38196.1 | |||
| BC139760.1 | ||||
| BC141341.1 |
More Information
Set Membership: |
|

Genomic Map