Assay Details
Target Gene Details
Entrez Gene ID: | 20170 |
Gene Name: | Hermansky-Pudlak syndrome 6 |
Gene Aliases: |
5330434M19Rik, BLOC2, ru |
Location: |
Chr.19:46003478-46006173 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hps6 | NM_176785.3 | 1 | 862 | NP_789742.2 |
| AF536239.1 | 1 | 807 | AAO25966.1 | |
| AK030582.1 | 1 | 833 | BAC27030.1 | |
| AK040849.1 | 1 | 809 | BAC30719.1 | |
| AK159588.1 | 1 | 828 | BAE35208.1 | |
| AK172058.1 | ||||
| BC034055.1 | 1 | 808 | AAH34055.1 |

Genomic Map