Assay Details
Target Gene Details
Entrez Gene ID: | 16580 |
Gene Name: | kinesin family member C5B |
Gene Aliases: |
Hset, KNSL2, Kifc5a |
Location: |
Chr.17:26917091-26932579 on Build GRCm38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Kifc5b | NM_053173.2 | NP_444403.2 | ||
| AK081484.1 | BAC38230.1 | |||
| BC091570.1 | 6 | 1760 | AAH91570.1 |
More Information
Set Membership: |
|

Genomic Map