Assay Details
Target Gene Details
Entrez Gene ID: | 19400 |
Gene Name: | receptor-associated protein of the synapse |
Gene Aliases: |
43kDa, Nraps, Raps, rapsyn |
Location: |
Chr.2:91035466-91045729 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rapsn | NM_009023.3 | NP_033049.2 | ||
| AK009672.1 | ||||
| BC111863.1 | AAI11864.1 | |||
| J03962.1 | AAA40030.1 | |||
| X15788.1 | CAA33789.1 |
More Information
Set Membership: |
|

Genomic Map