Assay Details
Target Gene Details
Entrez Gene ID: | 211556 |
Gene Name: | adaptor-related protein complex 1 associated regulatory protein |
Gene Aliases: |
AA407621, C78909, Gadkin |
Location: |
Chr.3:127807007-127837518 on Build GRCm38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ap1ar | NM_145964.2 | NP_666076.2 | ||
| AK037461.1 | 1 | 1459 | ||
| AK148810.1 | 2 | 3670 | ||
| BB611022.1 |
More Information
Set Membership: |
|

Genomic Map