Assay Details
Target Gene Details
Entrez Gene ID: | 15378 |
Gene Name: | hepatic nuclear factor 4, alpha |
Gene Aliases: |
HNF-4, Hnf4, Hnf4alpha, MODY1, Nr2a1, Tcf14 |
Location: |
Chr.2:163506809-163572907 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hnf4a | NM_008261.2 | NP_032287.2 | ||
| AF015275.1 | AAC02906.1 | |||
| AK143948.1 | BAE25624.1 | |||
| BC039220.1 | AAH39220.1 | |||
| D29015.1 | BAA06101.1 | |||
| EF193391.1 | ||||
| EF193393.1 | ||||
| FJ608819.1 | ||||
| FJ608820.1 | ||||
| FJ608821.1 |
More Information
Set Membership: |
|

Genomic Map