Assay Details
Target Gene Details
Entrez Gene ID: | 19013 |
Gene Name: | peroxisome proliferator activated receptor alpha |
Gene Aliases: |
4933429D07Rik, AW742785, Nr1c1, PPAR-alpha, PPARalpha, Ppar |
Location: |
Chr.15:85728192-85806851 on Build GRCm38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ppara | NM_001113418.1 | NP_001106889.1 | ||
| NM_011144.6 | NP_035274.2 | |||
| AK035676.1 | BAC29149.1 | |||
| AK081709.1 | BAC38303.1 | |||
| AK149460.1 | BAE28893.1 | |||
| BC016892.1 | AAH16892.1 | |||
| X57638.1 | CAA40856.1 |
More Information
Set Membership: |
|

Genomic Map