Assay Details
Target Gene Details
Entrez Gene ID: | 75019 |
Gene Name: | ribonuclease, RNase A family, 10 (non-active) |
Gene Aliases: |
4930474F22Rik, Rah1 |
Location: |
Chr.14:51007751-51010761 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rnase10 | NM_001162863.1 | NP_001156335.1 | ||
| NM_029145.3 | NP_083421.2 | |||
| AK015573.1 | BAB29898.1 | |||
| AY226990.1 | AAP43947.1 | |||
| BC141116.1 |
More Information
Set Membership: |
|

Genomic Map