Assay Details
Target Gene Details
Entrez Gene ID: | 68480 |
Gene Name: | RIKEN cDNA 1110007C09 gene |
Gene Aliases: |
AI851695, Bincard |
Location: |
Chr.13:49202950-49216067 on Build GRCm38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 1110007C09Rik | NM_026738.2 | NP_081014.1 | ||
| AK003526.1 | BAB22838.1 | |||
| AK154517.1 | BAE32646.1 | |||
| BC047205.1 | AAH47205.1 | |||
| BC147073.1 | ||||
| BC147074.1 |
More Information
Set Membership: |
|

Genomic Map