Assay Details
Target Gene Details
Entrez Gene ID: | 100504183 |
Gene Name: | keratin associated protein 16-1 |
Gene Aliases: |
Gm11570, OTTMUSG00000002209 |
Location: |
Chr.11:99984703-99986593 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Krtap16-1 | NM_001256057.1 | 1 | 192 | NP_001242986.1 |

Genomic Map